Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Instrument Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
Our Instagram
Our Facebook
Blog Behind the Bench
Customer Experience Center (CEC)
Ecommerce Exclusives
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CRH SIRNA Assays ›
Gene Symbol:
Crh
Gene Name:
corticotropin releasing hormone
Gene Aliases:
CRF
Chromosome Location:
Chr.2: 104459999 - 104461863 on Build Rnor_6.0
UniGene ID:
Rn.10349
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
194923
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Crh
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_031019.1
NP_112281.1
2
337
GenBank mRNA
X03036.1
-
2
335
Back To Top
More Information
Panther Classification:
Molecular Function -
peptide hormone
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of protein phosphorylation
synaptic transmission, dopaminergic
glucocorticoid biosynthetic process
inflammatory response
female pregnancy
learning or memory
steroid metabolic process
positive regulation of cell proliferation
associative learning
hormone-mediated apoptotic signaling pathway
positive regulation of gene expression
negative regulation of gene expression
negative regulation of norepinephrine secretion
positive regulation of cell death
regulation of serotonin secretion
diterpenoid metabolic process
hypothalamus development
lung development
adrenal gland development
positive regulation of cAMP biosynthetic process
negative regulation of epinephrine secretion
negative regulation of luteinizing hormone secretion
locomotory exploration behavior
positive regulation of insulin secretion involved in cellular response to glucose stimulus
response to immobilization stress
response to cocaine
response to drug
response to estrogen
response to ethanol
response to ether
negative regulation of blood pressure
response to pain
ion homeostasis
response to corticosterone
positive regulation of corticotropin secretion
positive regulation of cortisol secretion
long-term synaptic potentiation
positive regulation of digestive system process
negative regulation of cell death
negative regulation of glucagon secretion
cellular response to cocaine
cellular response to dexamethasone stimulus
positive regulation of calcium ion import
regulation of N-methyl-D-aspartate selective glutamate receptor activity
positive regulation of corticosterone secretion
positive regulation of behavioral fear response
hormone activity
protein binding
corticotropin-releasing hormone receptor 1 binding
corticotropin-releasing hormone receptor 2 binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0