Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Kcnj11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031358.3 | NP_112648.2 | 1 | 1381 | ||
XM_008759421.1 | XP_008757643.1 | 2 | 1074 | |||
GenBank mRNA | AB043638.1 | - | 1 | 1148 | ||
D61687.1 | - | 1 | 1389 | |||
U44897.1 | - | 1 | 1187 | |||
X97041.1 | - | 1 | 1145 |