Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Mybbp1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031668.1 | NP_113856.1 | 15 | 1883 | ||
XM_006246802.1 | XP_006246864.1 | 16 | 2127 | |||
GenBank mRNA | U83590.1 | - | 15 | 1883 |