Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Defb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031810.1 | NP_113998.1 | 2 | 162 | ||
GenBank mRNA | AF068860.1 | - | 2 | 138 | ||
AF093536.1 | - | 2 | 162 |