Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Gne | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053765.2 | NP_446217.1 | 2, 3 | 503 | ||
XM_006238006.2 | XP_006238068.1 | 2, 3 | 321 | |||
XM_006238008.1 | XP_006238070.1 | 2, 3 | 287 | |||
XM_006238009.2 | XP_006238071.1 | 2, 3 | 664 | |||
GenBank mRNA | BC062011.1 | - | 2, 3 | 503 | 5621384 | |
FQ224587.1 | - | 1, 2 | 166 | |||
Y07744.1 | - | 1, 2 | 211 |