Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ALDH1A2 SIRNA Assays ›
Gene Symbol:
Aldh1a2
Gene Name:
aldehyde dehydrogenase 1 family, member A2
Gene Aliases:
Raldh-2
Chromosome Location:
Chr.8: 77640234 - 77719488 on Build Rnor_6.0
UniGene ID:
Rn.10514
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
195819
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Aldh1a2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_053896.2
NP_446348.2
4
420
GenBank mRNA
BC098910.1
-
4
412
7455646
BE111477.1
-
4
420
U60063.1
-
4
416
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_009022, CJ166631, X99273, BC075704, AK078553.
Panther Classification:
Molecular Function -
dehydrogenase
oxidoreductase
metabolite interconversion enzyme
Gene Ontology Categories:
Function(s)
Process(es)
blood vessel development
kidney development
liver development
regulation of endothelial cell proliferation
retinoic acid biosynthetic process
heart morphogenesis
midgut development
positive regulation of cell proliferation
negative regulation of cell proliferation
determination of bilateral symmetry
anterior/posterior pattern specification
proximal/distal pattern formation
positive regulation of gene expression
neural crest cell development
morphogenesis of embryonic epithelium
neural tube development
pituitary gland development
neuron differentiation
lung development
embryonic limb morphogenesis
forebrain development
hindbrain development
pancreas development
embryonic camera-type eye development
response to estradiol
response to vitamin A
response to cytokine
embryonic forelimb morphogenesis
ureter maturation
retinol metabolic process
retinoic acid metabolic process
retinal metabolic process
9-cis-retinoic acid biosynthetic process
camera-type eye development
positive regulation of apoptotic process
retinoic acid receptor signaling pathway
embryonic digestive tract development
cardiac muscle tissue development
oxidation-reduction process
face development
cellular response to retinoic acid
retinal dehydrogenase activity
3-chloroallyl aldehyde dehydrogenase activity
aldehyde dehydrogenase (NAD) activity
retinal binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-chn84:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0