Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NME1 SIRNA Assays ›
Gene Symbol:
Nme1
Gene Name:
NME/NM23 nucleoside diphosphate kinase 1
Gene Aliases:
-
Chromosome Location:
Chr.10: 81657153 - 81666523 on Build Rnor_6.0
UniGene ID:
Rn.6236
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
196242
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Nme1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_138548.2
NP_612557.1
3
268
GenBank mRNA
D13374.1
-
3
269
Back To Top
More Information
Panther Classification:
Molecular Function -
kinase
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of myeloid leukocyte differentiation
purine nucleotide metabolic process
nucleoside diphosphate phosphorylation
GTP biosynthetic process
pyrimidine nucleotide metabolic process
UTP biosynthetic process
CTP biosynthetic process
DNA catabolic process
endocytosis
integrin-mediated signaling pathway
lactation
nucleoside triphosphate biosynthetic process
negative regulation of gene expression
positive regulation of neuron projection development
response to amine
hippocampus development
mammary gland development
response to testosterone
cellular response to drug
response to drug
regulation of apoptotic process
negative regulation of apoptotic process
positive regulation of DNA binding
positive regulation of keratinocyte differentiation
positive regulation of transcription from RNA polymerase II promoter
positive regulation of epithelial cell proliferation
response to cAMP
cellular response to glucose stimulus
cellular response to fatty acid
magnesium ion binding
RNA polymerase II regulatory region sequence-specific DNA binding
single-stranded DNA binding
deoxyribonuclease activity
nucleoside diphosphate kinase activity
protein binding
ATP binding
GTP binding
intermediate filament binding
enzyme binding
protein kinase binding
identical protein binding
gamma-tubulin binding
ribosomal small subunit binding
poly(A) RNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-mfv9s:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0