Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: GPR17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161415.1 | NP_001154887.1 | 4 | 2155 | ||
NM_001161416.1 | NP_001154888.1 | 3 | 2090 | |||
NM_001161417.1 | NP_001154889.1 | 2 | 1861 | |||
NM_005291.2 | NP_005282.1 | 3 | 1926 | |||
XM_017003833.1 | XP_016859322.1 | 2 | 2282 | |||
GenBank mRNA | AK126849.1 | - | 1 | 3116 | ||
BC031653.1 | - | 2 | 1540 | 5174146 | ||
BC039595.1 | - | 2 | 1545 | |||
Y12546.1 | - | 3 | 1618 | |||
Z94154.1 | - | 2 | 1554 | |||
Z94155.1 | - | 1 | 1338 |