Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BMP6 SIRNA Assays ›
Gene Symbol:
Bmp6
Gene Name:
bone morphogenetic protein 6
Gene Aliases:
VGR
Chromosome Location:
Chr.17: 26955142 - 27112820 on Build Rnor_6.0
UniGene ID:
Rn.40476
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
197766
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Bmp6
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_013107.1
NP_037239.1
2
794
GenBank mRNA
AY184240.1
-
2
794
U66298.1
-
2
567
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Human: NM_001718, BC128031, BC099624, M60315, BC113018, and others.
Panther Classification:
Molecular Function -
growth factor
Gene Ontology Categories:
Function(s)
Process(es)
ossification
osteoblast differentiation
eye development
kidney development
positive regulation of endothelial cell proliferation
endochondral ossification
type B pancreatic cell development
cellular iron ion homeostasis
inflammatory response
immune response
response to iron ion
positive regulation of pathway-restricted SMAD protein phosphorylation
response to activity
cell differentiation
positive regulation of bone mineralization
BMP signaling pathway
male genitalia development
response to magnesium ion
positive regulation of chondrocyte differentiation
positive regulation of aldosterone biosynthetic process
response to retinoic acid
growth
regulation of apoptotic process
regulation of MAPK cascade
regulation of cell differentiation
positive regulation of endothelial cell differentiation
positive regulation of neuron differentiation
positive regulation of osteoblast differentiation
positive regulation of transcription from RNA polymerase II promoter
cell development
positive regulation of epithelial cell proliferation
cartilage development
response to glucocorticoid
SMAD protein signal transduction
cellular response to mechanical stimulus
positive regulation of aldosterone secretion
cytokine activity
transforming growth factor beta receptor binding
growth factor activity
protein heterodimerization activity
BMP receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-n5w8t:80/100.66.78.251:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0