Search
Search

Chka| Gene Symbol: Chka | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017127.1 | NP_058823.1 | 2 | 672 | ||
| XM_006230707.2 | XP_006230769.1 | 2 | 1611 | |||
| XM_006230708.2 | XP_006230770.1 | 2 | 1610 | |||
| GenBank mRNA | BC081821.1 | - | 2 | 648 | 7109261 | |
| D10262.1 | - | 2 | 672 | |||
| D37885.1 | - | 2 | 374 | |||