Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RPS3A SIRNA Assays ›
          Gene Symbol:
          Rps3a
          Gene Name:
          ribosomal protein S3a
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 185440477 - 185444846 on Build Rnor_6.0
          UniGene ID:
          Rn.94935
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID 198156

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Rps3a
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_017153.1 NP_058849.1 4 468
          GenBank mRNA BC058483.1 - 4 473 6887884
          FQ209727.1 - 4 468
          FQ211699.1 - 4 469
          FQ211708.1 - 4 468
          FQ216850.1 - 4 468
          FQ217809.1 - 4 468
          FQ219020.1 - 4 468
          FQ219404.1 - 4 467
          FQ219522.1 - 4 469
          FQ219543.1 - 4 469
          FQ219867.1 - 4 469
          FQ220002.1 - 4 468
          FQ220027.1 - 4 468
          FQ220037.1 - 4 469
          FQ220045.1 - 4 468
          FQ220135.1 - 4 469
          FQ220260.1 - 4 468
          FQ220261.1 - 4 469
          FQ220317.1 - 4 468
          FQ220504.1 - 4 468
          FQ220577.1 - 4 469
          FQ220731.1 - 4 468
          FQ221431.1 - 4 469
          FQ221460.1 - 4 468
          FQ221744.1 - 4 468
          FQ221931.1 - 4 468
          FQ222056.1 - 4 468
          FQ222736.1 - 4 469
          FQ222773.1 - 4 469
          FQ222846.1 - 4 498
          FQ222869.1 - 4 468
          FQ222876.1 - 4 468
          FQ223535.1 - 4 475
          FQ223540.1 - 4 468
          FQ225080.1 - 4 469
          FQ225145.1 - 4 468
          FQ225713.1 - 4 467
          FQ227696.1 - 4 468
          FQ227805.1 - 4 469
          FQ227878.1 - 4 469
          FQ228312.1 - 4 469
          FQ228317.1 - 4 468
          FQ228376.1 - 4 469
          FQ228558.1 - 4 468
          FQ229234.1 - 4 468
          FQ229381.1 - 4 469
          FQ229520.1 - 4 468
          FQ229870.1 - 4 468
          FQ229960.1 - 4 468
          FQ231162.1 - 4 469
          FQ231673.1 - 4 468
          FQ232329.1 - 4 470
          FQ232836.1 - 4 468
          FQ232928.1 - 4 469
          FQ233768.1 - 4 472
          FQ234311.1 - 4 468
          FQ234537.1 - 4 468
          M84716.1 - 4 462
          X75161.1 - 4 468

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          ribosomal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          cytoplasmic translation
          translation
          biological_process
          cell differentiation
          negative regulation of apoptotic process
          molecular_function
          structural constituent of ribosome
          translation initiation factor binding
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0