Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HTR2A SIRNA Assays ›
Gene Symbol:
Htr2a
Gene Name:
5-hydroxytryptamine (serotonin) receptor 2A, G protein-coupled
Gene Aliases:
5-HT2A, 5Ht-2
Chromosome Location:
Chr.15: 56666152 - 56732469 on Build Rnor_6.0
UniGene ID:
Rn.10294
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
198396
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Htr2a
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_017254.1
NP_058950.1
3
860
GenBank mRNA
M30705.1
-
3
860
M64867.1
-
1266
X13971.1
-
1403
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_172812, BC108972, BC108973, AK053140, S49542.
Set Membership:
G-protein coupled receptor
Panther Classification:
Molecular Function -
G-protein coupled receptor
Gene Ontology Categories:
Function(s)
Process(es)
temperature homeostasis
cellular calcium ion homeostasis
G-protein coupled receptor signaling pathway
activation of phospholipase C activity
positive regulation of cytosolic calcium ion concentration
phospholipase C-activating serotonin receptor signaling pathway
serotonin receptor signaling pathway
aging
memory
cell death
positive regulation of cell proliferation
positive regulation of phosphatidylinositol biosynthetic process
regulation of dopamine secretion
phosphatidylinositol 3-kinase signaling
artery smooth muscle contraction
urinary bladder smooth muscle contraction
sensory perception of pain
sleep
positive regulation of kinase activity
response to drug
negative regulation of potassium ion transport
positive regulation of MAP kinase activity
protein localization to cytoskeleton
positive regulation of fat cell differentiation
positive regulation of glycolytic process
positive regulation of vasoconstriction
regulation of hormone secretion
behavioral response to cocaine
positive regulation of peptidyl-tyrosine phosphorylation
regulation of behavior
detection of temperature stimulus involved in sensory perception of pain
detection of mechanical stimulus involved in sensory perception of pain
release of sequestered calcium ion into cytosol
negative regulation of synaptic transmission, glutamatergic
positive regulation of ERK1 and ERK2 cascade
G-protein alpha-subunit binding
G-protein coupled serotonin receptor activity
protein binding
drug binding
protein complex binding
serotonin binding
1-(4-iodo-2,5-dimethoxyphenyl)propan-2-amine binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System