Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CALM2 SIRNA Assays ›
Gene Symbol:
Calm2
Gene Name:
calmodulin 2
Gene Aliases:
Cam
Chromosome Location:
Chr.6: 11064750 - 11080134 on Build Rnor_6.0
UniGene ID:
Rn.5968
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
198542
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Calm2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
GenBank mRNA
BC058485.1
-
1042
6921326
FQ211494.1
-
1042
FQ211970.1
-
1037
FQ212357.1
-
1039
FQ212579.1
-
1038
FQ212640.1
-
1038
FQ212691.1
-
1038
FQ212869.1
-
1042
FQ213028.1
-
1038
FQ213029.1
-
1036
FQ213115.1
-
1044
FQ213200.1
-
1041
FQ213216.1
-
1038
FQ213310.1
-
931
FQ213366.1
-
1037
FQ213496.1
-
1041
FQ213520.1
-
1042
FQ213526.1
-
1038
FQ213581.1
-
1038
FQ213598.1
-
1038
FQ213637.1
-
1038
FQ213695.1
-
1038
FQ213703.1
-
1038
FQ213760.1
-
1038
FQ213762.1
-
1043
FQ213913.1
-
1038
FQ213941.1
-
1042
FQ213981.1
-
1039
FQ213985.1
-
932
FQ213999.1
-
1037
FQ214059.1
-
1042
FQ214181.1
-
1042
FQ214262.1
-
1042
FQ214491.1
-
1038
FQ214500.1
-
1038
FQ214519.1
-
1038
FQ214525.1
-
1038
FQ214540.1
-
1042
FQ215159.1
-
1042
FQ215317.1
-
1038
FQ215600.1
-
1042
FQ216397.1
-
1038
FQ216471.1
-
1039
FQ218091.1
-
1038
FQ220250.1
-
1038
FQ220325.1
-
1038
FQ220698.1
-
1042
FQ220779.1
-
1038
FQ222184.1
-
1041
FQ223357.1
-
1042
FQ225272.1
-
1039
FQ226994.1
-
1042
FQ229228.1
-
1038
FQ229580.1
-
1042
FQ229619.1
-
1038
FQ229905.1
-
1037
FQ231022.1
-
1042
FQ231702.1
-
1042
FQ233337.1
-
1041
FQ234377.1
-
1042
FQ235266.1
-
1039
M17069.1
-
1039
M19312.1
-
1011
Back To Top
More Information
Panther Classification:
Molecular Function -
calmodulin-related
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0