Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CCR5 SIRNA Assays ›
          Gene Symbol:
          CCR5
          Gene Name:
          C-C motif chemokine receptor 5 (gene/pseudogene)
          Gene Aliases:
          CC-CKR-5, CCCKR5, CCR-5, CD195, CKR-5, CKR5, CMKBR5, IDDM22
          Chromosome Location:
          Chr.3: 46370142 - 46376206 on Build GRCh38
          UniGene ID:
          Hs.450802
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 1987

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CCR5
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000579.3 NP_000570.1 3 544
          NM_001100168.1 NP_001093638.1 3 309
          GenBank mRNA AF011500.1 - 1 191
          AF011501.1 - 1 191
          AF011502.1 - 1 191
          AF011503.1 - 1 191
          AF011504.1 - 1 190
          AF011505.1 - 1 191
          AF011506.1 - 1 191
          AF011507.1 - 1 191
          AF011508.1 - 1 191
          AF011509.1 - 1 191
          AF011510.1 - 1 191
          AF011511.1 - 1 191
          AF011512.1 - 1 191
          AF011513.1 - 1 191
          AF011514.1 - 1 191
          AF011515.1 - 1 191
          AF011516.1 - 1 191
          AF011517.1 - 1 191
          AF011518.1 - 1 191
          AF011519.1 - 1 191
          AF011520.1 - 1 191
          AF011521.1 - 1 191
          AF011522.1 - 1 191
          AF011523.1 - 1 191
          AF011524.1 - 1 191
          AF011525.1 - 1 191
          AF011527.1 - 1 191
          AF011528.1 - 1 191
          AF011529.1 - 1 191
          AF011531.1 - 1 191
          AF011532.1 - 1 191
          AF011533.1 - 1 191
          AF011534.1 - 1 191
          AF011535.1 - 1 191
          AF011536.1 - 1 191
          AF011537.1 - 1 191
          AK292132.1 - 2 241
          AK314882.1 - 2 333
          BC038398.1 - 2 230 5186388
          DA818906.1 - 2 333
          U54994.1 - 2 241
          U57840.1 - 2 213

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: BC105643, AK170303.

          Set Membership:

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          dendritic cell chemotaxis
          calcium ion transport
          chemotaxis
          inflammatory response
          immune response
          cellular defense response
          cell surface receptor signaling pathway
          G-protein coupled receptor signaling pathway
          positive regulation of cytosolic calcium ion concentration
          cell-cell signaling
          release of sequestered calcium ion into cytosol by sarcoplasmic reticulum
          fusion of virus membrane with host plasma membrane
          calcium-mediated signaling
          signaling
          entry into host cell
          chemokine-mediated signaling pathway
          response to cholesterol
          cellular response to lipopolysaccharide
          virus receptor activity
          actin binding
          phosphatidylinositol phospholipase C activity
          chemokine receptor activity
          protein binding
          coreceptor activity
          C-C chemokine receptor activity
          C-C chemokine binding
          chemokine (C-C motif) ligand 5 binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0