Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Phgdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031620.1 | NP_113808.1 | 7, 8 | 760 | ||
XM_006233010.1 | XP_006233072.1 | 6, 7 | 783 | |||
GenBank mRNA | BC086327.1 | - | 6, 7 | 788 | 7115765 | |
X97772.1 | - | 6, 7 | 719 |