Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FBXW11 SIRNA Assays ›
          Gene Symbol:
          FBXW11
          Gene Name:
          F-box and WD repeat domain containing 11
          Gene Aliases:
          BTRC2, BTRCP2, FBW1B, FBXW1B, Fbw11, Hos
          Chromosome Location:
          Chr.5: 171861549 - 172006873 on Build GRCh38
          UniGene ID:
          Hs.484138
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 19965

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 286.00
          Your Price
          Online offer: 277.65
          277.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: FBXW11
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_012300.2 NP_036432.2 4 758
          NM_033644.2 NP_387448.2 4 719
          NM_033645.2 NP_387449.2 3 656
          XM_005265855.4 XP_005265912.1 5 808
          XM_005265856.2 XP_005265913.1 6 778
          XM_005265857.2 XP_005265914.1 3 316
          XM_005265858.2 XP_005265915.1 5 820
          XM_017009279.1 XP_016864768.1 4 618
          XM_017009280.1 XP_016864769.1 5 634
          XM_017009281.1 XP_016864770.1 5 702
          GenBank mRNA AB014596.1 - 4 412
          AB033279.1 - 3 386
          AB033280.1 - 4 504
          AB033281.1 - 4 526
          AF176022.1 - 4 411
          AK294956.1 - 4 401
          AK314999.1 - 3 285
          BC026213.1 - 4 719 4419029

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001106993, XM_006246092, XM_006246093, XM_006246094.
          This siRNA also targets the following transcripts in Mouse: NM_001271347, NM_001271349, NM_134015, NM_001271348, AK168667, and others.

          Gene Ontology Categories:

          Function(s) Process(es)

          G2/M transition of mitotic cell cycle
          protein polyubiquitination
          stimulatory C-type lectin receptor signaling pathway
          protein dephosphorylation
          Wnt signaling pathway
          protein ubiquitination
          SCF-dependent proteasomal ubiquitin-dependent protein catabolic process
          protein destabilization
          NIK/NF-kappaB signaling
          Fc-epsilon receptor signaling pathway
          negative regulation of NF-kappaB import into nucleus
          positive regulation of circadian rhythm
          proteasome-mediated ubiquitin-dependent protein catabolic process
          positive regulation of proteolysis
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          rhythmic process
          T cell receptor signaling pathway
          stress-activated MAPK cascade
          ubiquitin-protein transferase activity
          protein binding
          protein dimerization activity
          ubiquitin protein ligase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0