Search
Search

Dnajb9| Gene Symbol: Dnajb9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012699.2 | NP_036831.2 | 2 | 283 | ||
| XM_006240065.2 | XP_006240127.1 | 2 | 274 | |||
| GenBank mRNA | BC070915.1 | - | 2 | 281 | 7104653 | |
| FQ230361.1 | - | 1 | 36 | |||
| X98993.2 | - | 2 | 283 | |||