Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Clinical Genomics
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CD24 SIRNA Assays ›
Gene Symbol:
Cd24
Gene Name:
CD24 molecule
Gene Aliases:
Cd24a
Chromosome Location:
Chr.20: 48335540 - 48340847 on Build Rnor_6.0
UniGene ID:
Rn.6007
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
200359
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Cd24
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_012752.3
NP_036884.1
2
554
GenBank mRNA
BC064439.1
-
2
538
5623514
BC081851.1
-
2
549
7110912
U49062.1
-
2
538
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
response to hypoxia
cell activation
regulation of cytokine-mediated signaling pathway
response to molecule of bacterial origin
pre-B cell differentiation
leukocyte migration involved in inflammatory response
positive regulation of T cell mediated immune response to tumor cell
positive regulation of inflammatory response to antigenic stimulus
positive regulation of B cell apoptotic process
immune response
cell adhesion
heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules
leukocyte cell-cell adhesion
cell surface receptor signaling pathway
positive regulation of cytosolic calcium ion concentration
cell-cell signaling
neuromuscular synaptic transmission
negative regulation of neuroblast proliferation
apoptotic mitochondrial changes
positive regulation of neuron projection development
single organismal cell-cell adhesion
cell migration
regulation of cell-cell adhesion
positive regulation of cell-cell adhesion
apoptotic nuclear changes
negative regulation of B cell proliferation
neuron projection development
T cell costimulation
B cell receptor transport into membrane raft
chemokine receptor transport out of membrane raft
negative regulation of transforming growth factor beta3 production
positive regulation of integrin activation
positive regulation of cell adhesion mediated by integrin
regulation of cell-cell adhesion mediated by integrin
positive regulation of cell-cell adhesion mediated by integrin
negative regulation of erythrocyte clearance
homotypic cell-cell adhesion
negative regulation of erythrocyte aggregation
positive regulation of T cell homeostatic proliferation
positive regulation of activated T cell proliferation
regulation of phosphorylation
cholesterol homeostasis
positive regulation of MAP kinase activity
regulation of MAPK cascade
response to estrogen
regulation of B cell differentiation
negative regulation of neuron differentiation
respiratory burst
negative regulation of T cell homeostatic proliferation
positive regulation of alpha-beta T cell proliferation
synaptic vesicle endocytosis
positive regulation of peptidyl-tyrosine phosphorylation
negative regulation of neurogenesis
positive regulation of calcium-mediated signaling
positive regulation of protein tyrosine kinase activity
glomerular visceral epithelial cell differentiation
glomerular parietal epithelial cell differentiation
apoptotic signaling pathway
positive regulation of nephron tubule epithelial cell differentiation
protein kinase binding
carbohydrate binding
protein tyrosine kinase activator activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-7l2qm:80/100.66.75.127:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline