Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other EPO SIRNA Assays ›
Gene Symbol:
Epo
Gene Name:
erythropoietin
Gene Aliases:
-
Chromosome Location:
Chr.12: 22274828 - 22278268 on Build Rnor_6.0
UniGene ID:
Rn.11365
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
200565
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Epo
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_017001.1
NP_058697.1
1378
GenBank mRNA
D10763.1
-
1378
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
response to hypoxia
regulation of transcription from RNA polymerase II promoter
apoptotic process
embryo implantation
aging
response to nutrient
positive regulation of cell proliferation
response to salt stress
negative regulation of calcium ion transport into cytosol
peptidyl-serine phosphorylation
erythrocyte differentiation
response to lipopolysaccharide
negative regulation of myeloid cell apoptotic process
response to vitamin A
response to testosterone
positive regulation of tyrosine phosphorylation of Stat5 protein
hemoglobin biosynthetic process
negative regulation of apoptotic process
erythrocyte maturation
response to estrogen
positive regulation of neuron differentiation
positive regulation of DNA replication
positive regulation of protein kinase activity
positive regulation of transcription, DNA-templated
positive regulation of Ras protein signal transduction
response to axon injury
response to electrical stimulus
response to hyperoxia
response to interleukin-1
cellular hyperosmotic response
negative regulation of intrinsic apoptotic signaling pathway in response to osmotic stress
negative regulation of erythrocyte apoptotic process
negative regulation of cation channel activity
erythropoietin receptor binding
hormone activity
protein kinase activator activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0