Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PTH SIRNA Assays ›
Gene Symbol:
Pth
Gene Name:
parathyroid hormone
Gene Aliases:
PTH-(1-84), Pth1, Pthr1
Chromosome Location:
Chr.1: 178215829 - 178218761 on Build Rnor_6.0
UniGene ID:
Rn.9768
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
200675
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Pth
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_017044.1
NP_058740.1
3
220
GenBank mRNA
S80127.1
-
3
189
X05721.1
-
3
220
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_020623, BC099456, AK138677.
Panther Classification:
Molecular Function -
peptide hormone
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
skeletal system development
ossification
transcription from RNA polymerase II promoter
cellular calcium ion homeostasis
adenylate cyclase-activating G-protein coupled receptor signaling pathway
Rho protein signal transduction
cell-cell signaling
positive regulation of signal transduction
response to lead ion
regulation of gene expression
positive regulation of bone mineralization
positive regulation of cAMP biosynthetic process
response to nutrient levels
response to vitamin D
cellular macromolecule biosynthetic process
response to drug
bone resorption
response to ethanol
positive regulation of glycogen biosynthetic process
positive regulation of ossification
positive regulation of transcription from RNA polymerase II promoter
positive regulation of glucose import
response to cadmium ion
response to parathyroid hormone
response to fibroblast growth factor
transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
hormone activity
parathyroid hormone receptor binding
type 1 parathyroid hormone receptor binding
peptide hormone receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0