Search Thermo Fisher Scientific
Gene Symbol: Tbxa2r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017054.1 | NP_058750.1 | 3 | 1817 | ||
XM_006240846.1 | XP_006240908.1 | 3 | 1989 | |||
XM_006240847.1 | XP_006240909.1 | 2 | 1541 | |||
GenBank mRNA | D32080.1 | - | 3 | 1817 |
Set Membership: |
G-protein coupled receptor |