Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Tnfrsf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019135.1 | NP_062008.1 | 5 | 495 | ||
XM_008764206.1 | XP_008762428.1 | 5 | 1059 | |||
GenBank mRNA | D42117.1 | - | 5 | 495 |