Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Enterprise Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other GPX1 SIRNA Assays ›
          Gene Symbol:
          Gpx1
          Gene Name:
          glutathione peroxidase 1
          Gene Aliases:
          GSHPx, GSHPx-1
          Chromosome Location:
          Chr.8: 117117430 - 117118528 on Build Rnor_6.0
          UniGene ID:
          Rn.11323
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID 201043

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Gpx1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_030826.3 NP_110453.3 2 542
          GenBank mRNA BC058438.1 - 2 514 6918849
          CB579218.1 - 2 542
          FQ211052.1 - 2 539
          FQ227523.1 - 2 571
          M21210.1 - 2 548

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          peroxidase

          Gene Ontology Categories:

          Function(s) Process(es)

          response to reactive oxygen species
          temperature homeostasis
          endothelial cell development
          negative regulation of inflammatory response to antigenic stimulus
          lipid metabolic process
          triglyceride metabolic process
          glutathione metabolic process
          apoptotic process
          response to oxidative stress
          response to lipid hydroperoxide
          aging
          sensory perception of sound
          cell proliferation
          intrinsic apoptotic signaling pathway in response to oxidative stress
          response to xenobiotic stimulus
          response to symbiotic bacterium
          response to wounding
          response to toxic substance
          UV protection
          response to hormone
          response to glucose
          response to selenium ion
          response to gamma radiation
          response to organic cyclic compound
          myotube differentiation
          protein oxidation
          response to estradiol
          response to hydroperoxide
          regulation of mammary gland epithelial cell proliferation
          response to nicotine
          regulation of gene expression, epigenetic
          vasodilation
          response to hydrogen peroxide
          hydrogen peroxide catabolic process
          negative regulation of apoptotic process
          negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
          skeletal muscle tissue regeneration
          regulation of neuron apoptotic process
          blood vessel endothelial cell migration
          fat cell differentiation
          cell redox homeostasis
          skeletal muscle fiber development
          myoblast proliferation
          response to folic acid
          interaction with symbiont
          positive regulation of protein kinase B signaling
          heart contraction
          angiogenesis involved in wound healing
          regulation of proteasomal protein catabolic process
          negative regulation of release of cytochrome c from mitochondria
          negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
          negative regulation of oxidative stress-induced intrinsic apoptotic signaling pathway
          glutathione peroxidase activity
          selenium binding
          SH3 domain binding
          glutathione binding
          phospholipid-hydroperoxide glutathione peroxidase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0