Search
Search

Gnb1| Gene Symbol: Gnb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006239517.2 | XP_006239579.1 | 12 | 1702 | ||
| XM_006239518.2 | XP_006239580.1 | 12 | 1705 | |||
| XM_006239519.2 | XP_006239581.1 | 12 | 1701 | |||
| XM_006239520.2 | XP_006239582.1 | 12 | 1698 | |||
| XM_008764335.1 | XP_008762557.1 | 11 | 1585 | |||
| GenBank mRNA | AF022083.1 | - | 2 | 490 | ||
| U88324.1 | - | 10 | 1437 | |||