Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CIB1 SIRNA Assays ›
Gene Symbol:
Cib1
Gene Name:
calcium and integrin binding 1 (calmyrin)
Gene Aliases:
Cib, Sip2-28
Chromosome Location:
Chr.1: 142014962 - 142020461 on Build Rnor_6.0
UniGene ID:
Rn.833
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
201077
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Cib1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_031145.1
NP_112407.1
7
656
GenBank mRNA
AF136585.1
-
7
656
BC091143.1
-
7
642
7308828
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
angiogenesis
negative regulation of protein phosphorylation
positive regulation of protein phosphorylation
positive regulation of cell-matrix adhesion
response to ischemia
apoptotic process
cellular response to DNA damage stimulus
negative regulation of microtubule depolymerization
endomitotic cell cycle
cell adhesion
spermatid development
positive regulation of cell proliferation
negative regulation of cell proliferation
negative regulation of neuron projection development
platelet formation
positive regulation of cell growth
positive regulation of cell migration
cytoplasmic microtubule organization
positive regulation of cell adhesion mediated by integrin
thrombopoietin-mediated signaling pathway
regulation of cell proliferation
negative regulation of apoptotic process
positive regulation of catalytic activity
negative regulation of megakaryocyte differentiation
positive regulation of NF-kappaB transcription factor activity
cell division
regulation of cell division
negative regulation of protein kinase B signaling
positive regulation of ERK1 and ERK2 cascade
positive regulation of calcineurin-NFAT signaling cascade
cellular response to tumor necrosis factor
cellular response to growth factor stimulus
positive regulation of protein serine/threonine kinase activity
positive regulation of establishment of protein localization to plasma membrane
positive regulation of cell migration involved in sprouting angiogenesis
positive regulation of protein targeting to membrane
positive regulation of substrate adhesion-dependent cell spreading
cellular response to nerve growth factor stimulus
positive regulation of male germ cell proliferation
calcium ion binding
protein binding
Ras GTPase binding
protein anchor
ion channel binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-qvstf:80/100.66.73.196:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0