Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other DBI SIRNA Assays ›
          Gene Symbol:
          Dbi
          Gene Name:
          diazepam binding inhibitor (GABA receptor modulator, acyl-CoA binding protein)
          Gene Aliases:
          Acbp, Acoabp3, Ep, Odn, RNACOABP3, Ttn
          Chromosome Location:
          Chr.13: 36147667 - 36156076 on Build Rnor_6.0
          UniGene ID:
          Rn.3285
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID 201190

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Dbi
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_031853.4 NP_114054.1 4 343
          GenBank mRNA BC084717.1 - 3 230 7310272
          FQ209769.1 - 4 269
          FQ209835.1 - 4 291
          FQ209938.1 - 4 286
          FQ210140.1 - 4 291
          FQ210336.1 - 4 288
          FQ210460.1 - 4 291
          FQ210781.1 - 4 318
          FQ210997.1 - 4 288
          FQ211081.1 - 4 291
          FQ211136.1 - 4 291
          FQ211192.1 - 4 291
          FQ211339.1 - 4 292
          FQ211377.1 - 3 212
          FQ213527.1 - 4 292
          FQ214572.1 - 4 291
          FQ217714.1 - 4 292
          FQ218148.1 - 4 286
          FQ218164.1 - 4 310
          FQ218239.1 - 4 291
          FQ218300.1 - 4 291
          FQ218680.1 - 4 291
          FQ219136.1 - 4 291
          FQ219739.1 - 4 291
          FQ221153.1 - 4 286
          FQ221871.1 - 4 292
          FQ222052.1 - 4 291
          FQ222378.1 - 4 293
          FQ222416.1 - 4 286
          FQ222549.1 - 4 281
          FQ222564.1 - 4 292
          FQ223167.1 - 4 286
          FQ223413.1 - 4 291
          FQ223492.1 - 4 292
          FQ223501.1 - 4 291
          FQ223602.1 - 4 249
          FQ224361.1 - 4 291
          FQ224797.1 - 4 292
          FQ229114.1 - 4 286
          M14201.1 - 4 343
          M20268.1 - 4 232

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_007830, NM_001037999, X61431, AK008576, EF437365, and others.

          Panther Classification:

          Molecular Function -

          transfer/carrier protein

          Gene Ontology Categories:

          Function(s) Process(es)

          behavioral fear response
          hair follicle development
          acyl-CoA metabolic process
          triglyceride metabolic process
          steroid biosynthetic process
          brain development
          learning or memory
          glial cell proliferation
          protein palmitoylation
          lateral ventricle development
          pancreatic juice secretion
          cellular response to nutrient
          negative regulation of fatty acid beta-oxidation
          regulation of synaptic transmission, GABAergic
          phosphatidylcholine acyl-chain remodeling
          skin development
          positive regulation of lipid biosynthetic process
          positive regulation of release of sequestered calcium ion into cytosol
          long-term synaptic potentiation
          fatty-acyl-CoA binding
          protein binding
          lipid binding
          benzodiazepine receptor binding
          long-chain fatty acyl-CoA binding
          protein dimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-xmktn:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0