Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ALB SIRNA Assays ›
Gene Symbol:
Alb
Gene Name:
albumin
Gene Aliases:
Alb1, Albza
Chromosome Location:
Chr.14: 19176275 - 19191793 on Build Rnor_6.0
UniGene ID:
Rn.202968
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
201389
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Alb
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_134326.2
NP_599153.2
10
1245
GenBank mRNA
BC085359.1
-
10
1245
7303856
FQ209382.1
-
6
693
FQ209565.1
-
3
278
FQ209580.1
-
3
230
FQ209595.1
-
2
173
FQ209635.1
-
4
456
FQ209651.1
-
4
485
FQ209693.1
-
3
280
FQ209695.1
-
6
679
FQ209713.1
-
2
155
FQ209714.1
-
3
348
FQ209721.1
-
4
466
FQ209750.1
-
1
24
FQ209825.1
-
3
271
FQ209907.1
-
6
714
FQ209910.1
-
1
18
FQ209930.1
-
1
17
FQ209957.1
-
5
626
FQ209968.1
-
3
273
FQ210022.1
-
4
468
FQ210057.1
-
6
644
FQ210064.1
-
2
137
FQ210111.1
-
4
497
FQ210134.1
-
2
60
FQ210171.1
-
4
466
FQ210205.1
-
2
158
FQ210297.1
-
3
313
FQ210310.1
-
8
917
FQ210316.1
-
4
466
FQ210361.1
-
3
386
FQ210399.1
-
3
211
FQ210406.1
-
4
445
FQ210411.1
-
3
377
FQ210445.1
-
2
151
FQ210540.1
-
3
195
FQ210546.1
-
4
493
FQ210564.1
-
3
230
FQ210585.1
-
2
101
FQ210640.1
-
4
450
FQ210749.1
-
2
169
FQ210803.1
-
6
717
FQ210850.1
-
5
561
FQ210851.1
-
5
592
FQ210858.1
-
7
800
FQ210872.1
-
1
50
FQ210877.1
-
6
660
FQ210926.1
-
2
63
FQ210946.1
-
2
154
FQ210954.1
-
2
233
FQ210973.1
-
1
24
FQ210995.1
-
4
455
FQ211001.1
-
3
285
FQ211010.1
-
5
603
FQ211021.1
-
1
31
FQ211036.1
-
4
498
FQ211096.1
-
4
466
FQ211119.1
-
2
155
FQ211129.1
-
4
439
FQ211131.1
-
1
30
FQ211157.1
-
2
109
FQ211196.1
-
4
426
FQ211226.1
-
2
148
FQ211255.1
-
6
693
FQ211259.1
-
6
702
FQ211320.1
-
4
497
FQ211355.1
-
3
290
FQ211376.1
-
2
149
FQ211378.1
-
4
466
FQ211445.1
-
3
370
FQ211464.1
-
2
70
FQ218081.1
-
3
274
FQ218137.1
-
4
470
FQ218216.1
-
5
569
FQ218268.1
-
2
166
FQ218283.1
-
3
363
FQ218304.1
-
2
110
FQ218310.1
-
2
144
FQ218331.1
-
2
144
FQ218336.1
-
2
74
FQ218371.1
-
2
176
FQ218379.1
-
2
152
FQ218412.1
-
3
277
FQ218458.1
-
6
709
FQ218497.1
-
2
137
FQ218541.1
-
2
107
FQ218566.1
-
3
376
FQ218682.1
-
4
434
FQ218739.1
-
5
614
FQ218757.1
-
2
155
FQ218799.1
-
3
199
FQ218806.1
-
2
151
FQ218815.1
-
5
579
FQ218867.1
-
3
395
FQ218944.1
-
3
308
FQ218989.1
-
3
299
FQ219017.1
-
3
250
FQ219210.1
-
6
664
FQ219231.1
-
3
291
FQ219260.1
-
4
435
FQ219293.1
-
6
693
FQ219382.1
-
3
380
FQ219442.1
-
4
463
FQ219470.1
-
5
564
FQ219477.1
-
3
306
FQ219514.1
-
4
492
FQ219650.1
-
4
462
FQ219670.1
-
6
743
FQ219679.1
-
4
455
FQ219682.1
-
4
437
FQ219706.1
-
2
175
FQ219734.1
-
8
896
FQ219775.1
-
3
349
FQ224096.1
-
3
349
V01222.1
-
10
1230
Back To Top
More Information
Panther Classification:
Molecular Function -
transfer/carrier protein
Gene Ontology Categories:
Function(s)
Process(es)
retina homeostasis
transport
response to nutrient
cellular response to starvation
response to organic substance
hemolysis by symbiont of host erythrocytes
vasodilation
negative regulation of apoptotic process
positive regulation of circadian sleep/wake cycle, non-REM sleep
response to mercury ion
maintenance of mitochondrion location
response to platinum ion
DNA binding
fatty acid binding
drug binding
zinc ion binding
toxic substance binding
oxygen binding
enzyme binding
pyridoxal phosphate binding
chaperone binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-gpcss:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0