Search Thermo Fisher Scientific
Gene Symbol: TIMM9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304485.1 | NP_001291414.1 | 5 | 549 | ||
NM_001304486.1 | NP_001291415.1 | 4 | 427 | |||
NM_001304487.1 | NP_001291416.1 | 4 | 461 | |||
NM_012460.3 | NP_036592.1 | 6 | 738 | |||
NR_130750.1 | 5 | 673 | ||||
NR_130752.1 | 3 | 362 | ||||
NR_130753.1 | 3 | 354 | ||||
GenBank mRNA | AF150100.1 | - | 5 | 429 | ||
AF152353.1 | - | 3 | 212 | |||
AK000666.1 | - | 5 | 447 | |||
AK312095.1 | - | 4 | 373 | |||
BC020213.1 | - | 6 | 638 | 4640518 | ||
BC054875.1 | - | 4 | 351 | 6663143 | ||
BF034731.1 | - | 5 | 393 | |||
BM811095.1 | - | 5 | 585 | |||
DB110327.1 | - | 3 | 291 | |||
HY139570.1 | - | 3 | 276 |