Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NME2 SIRNA Assays ›
          Gene Symbol:
          NME2 NME1-NME2
          Gene Name:
          NME/NM23 nucleoside diphosphate kinase 2
          NME1-NME2 readthrough
          Gene Aliases:
          NDKB, NDPK-B, NDPKB, NM23-H2, NM23B, PUF
          NM23-LV, NMELV
          Chromosome Location:
          Chr.17: 51165435 - 51171744 on Build GRCh38
          Chromosome Location:
          Chr.17: 51153536 - 51171744 on Build GRCh38
          UniGene ID:
          Hs.463456
          Hs.463456
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID 202046

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NME2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001018137.2 NP_001018147.1 5 774
          NM_001018138.1 NP_001018148.1 4 632
          NM_001018139.2 NP_001018149.1 5 590
          NM_001198682.1 NP_001185611.1 4 640
          NM_002512.3 NP_002503.1 5 753
          GenBank mRNA BC002476.2 - 5 555 3349768
          BC095458.1 - 5 564 30524221
          BM556410.1 - 5 590
          CN267160.1 - 4 495
          EU794639.1 - 5 572
          L16785.1 - 5 572
          M36981.1 - 5 595
          X58965.1 - 5 595
          Gene Symbol: NME1-NME2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001018136.2 NP_001018146.1 8 999
          NR_037149.1 9 1219
          GenBank mRNA BC107894.1 - 9 1121 6717551
          DQ109675.1 - 8 919

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          purine nucleotide metabolic process
          nucleoside diphosphate phosphorylation
          GTP biosynthetic process
          pyrimidine nucleotide metabolic process
          UTP biosynthetic process
          CTP biosynthetic process
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          cell adhesion
          integrin-mediated signaling pathway
          nucleoside triphosphate biosynthetic process
          nucleobase-containing small molecule interconversion
          peptidyl-histidine phosphorylation
          negative regulation of apoptotic process
          positive regulation of keratinocyte differentiation
          regulation of epidermis development
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of epithelial cell proliferation
          mitophagy in response to mitochondrial depolarization
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          nucleoside diphosphate kinase activity
          protein histidine kinase activity
          protein binding
          ATP binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0