Search
Search

GPR22| Gene Symbol: GPR22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005295.2 | NP_005286.2 | 3 | 1801 | ||
| XM_011516056.2 | XP_011514358.1 | 2 | 1487 | |||
| GenBank mRNA | AB209270.1 | - | 1 | 711 | ||
| AK122621.1 | - | 3 | 1798 | |||
| AK307393.1 | - | 3 | 1798 | |||
| BC107128.2 | - | 1 | 455 | 40024013 | ||
| BX647649.1 | - | 2 | 665 | |||
| HQ995527.1 | - | 1 | 455 | |||
Set Membership: |
|