Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other VRK2 SIRNA Assays ›
          Gene Symbol:
          VRK2
          Gene Name:
          vaccinia related kinase 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 57907651 - 58163991 on Build GRCh38
          UniGene ID:
          Hs.715298
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 202110

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: VRK2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001130480.2 NP_001123952.1 8, 9 862
          NM_001130481.2 NP_001123953.1 8, 9 704
          NM_001130482.2 NP_001123954.1 8, 9 808
          NM_001130483.2 NP_001123955.1 8, 9 862
          NM_001288836.1 NP_001275765.1 9, 10 999
          NM_001288837.1 NP_001275766.1 11, 12 1422
          NM_001288838.1 NP_001275767.1 10, 11 1304
          NM_001288839.1 NP_001275768.1 7, 8 839
          NM_006296.6 NP_006287.2 8, 9 909
          XM_005264540.4 XP_005264597.1 8, 9 922
          XM_006712090.3 XP_006712153.1 8, 9 717
          XM_006712091.3 XP_006712154.1 8, 9 957
          XM_006712092.3 XP_006712155.1 9, 10 1094
          XM_006712093.3 XP_006712156.1 6, 7 799
          XM_011533092.2 XP_011531394.1 8, 9 706
          XM_017004858.1 XP_016860347.1 8, 9 862
          XM_017004859.1 XP_016860348.1 9, 10 1584
          XM_017004860.1 XP_016860349.1 8, 9 1514
          XM_017004861.1 XP_016860350.1 6, 7 613
          XM_017004862.1 XP_016860351.1 7, 8 1426
          XM_017004863.1 XP_016860352.1 8, 9 1006
          XM_017004864.1 XP_016860353.1 9, 10 1094
          XM_017004865.1 XP_016860354.1 9, 10 908
          XM_017004866.1 XP_016860355.1 9, 10 957
          GenBank mRNA AB000450.1 - 8, 9 807
          AJ512204.2 - 8, 9 807
          AK058199.1 - 8, 9 764
          AK223540.1 - 8, 9 764
          AK292727.1 - 10, 11 1290
          AK295193.1 - 8, 9 754
          AK296611.1 - 9, 10 984
          AY228367.1 - 8, 9 704
          AY228368.1 - 8, 9 839
          AY228369.1 - 8, 9 737
          AY228370.1 - 9, 10 929
          AY228371.1 - 7, 8 769
          AY228372.1 - 8, 9 792
          BC021663.1 - 8, 9 735
          BC027854.1 - 8, 9 860 5221315

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          regulation of cell shape
          viral process
          peptidyl-serine phosphorylation
          cellular response to oxidative stress
          regulation of MAPK cascade
          protein autophosphorylation
          regulation of interleukin-1-mediated signaling pathway
          protein serine/threonine kinase activity
          protein binding
          ATP binding
          protein kinase binding
          protein domain specific binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-9tlvb:80/100.66.74.202:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0