Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: FUK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145059.2 | NP_659496.2 | 11 | 1040 | ||
XM_006721162.1 | XP_006721225.1 | 10 | 970 | |||
XM_011522928.1 | XP_011521230.1 | 11 | 1218 | |||
XM_011522929.1 | XP_011521231.1 | 11 | 1083 | |||
XM_011522930.2 | XP_011521232.1 | 11 | 1051 | |||
XM_011522931.2 | XP_011521233.1 | 11 | 1051 | |||
XM_017023012.1 | XP_016878501.1 | 11 | 1067 | |||
XM_017023013.1 | XP_016878502.1 | 8 | 863 | |||
XM_017023014.1 | XP_016878503.1 | 6 | 995 | |||
GenBank mRNA | AJ441184.1 | - | 11 | 1039 | ||
AK056456.1 | - | 11 | 1039 | |||
AK074168.1 | - | 9 | 1285 | |||
AK092025.1 | - | 3 | 184 | |||
AK096727.1 | - | 3 | 246 | |||
AK128387.1 | - | 11 | 1136 | |||
AK315590.1 | - | 11 | 1043 | |||
BC013735.1 | - | 9 | 826 | |||
BC032542.1 | - | 11 | 1046 | 5534376 |