Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: HIGD1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099668.1 | NP_001093138.1 | 4 | 614 | ||
NM_001099669.1 | NP_001093139.1 | 4 | 467 | |||
NM_014056.3 | NP_054775.2 | 4 | 443 | |||
GenBank mRNA | AF077034.1 | - | 4 | 395 | ||
AF145385.1 | - | 4 | 382 | |||
AK225461.1 | - | 4 | 367 | |||
AK225571.1 | - | 4 | 398 | |||
AL110233.1 | - | 4 | 413 | |||
BC000601.1 | - | 4 | 381 | 3346355 | ||
BC009583.1 | - | 4 | 374 | 4093421 | ||
BC009594.1 | - | 4 | 380 | 4109190 | ||
BC070277.1 | - | 4 | 396 | 4686431 | ||
BG708126.1 | - | 4 | 448 |