Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: LINC00339 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_023918.2 | 2 | 832 | |||
NR_023919.2 | 2 | 790 | ||||
NR_109759.1 | 2 | 786 | ||||
NR_109760.1 | 2 | 787 | ||||
NR_109761.1 | 3 | 907 | ||||
NR_109762.1 | 3 | 911 | ||||
GenBank mRNA | AF161506.1 | - | 2 | 761 | ||
BC016053.1 | - | 2 | 745 | |||
BC056896.1 | - | 1 | 390 |