Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TUBGCP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286414.2 | NP_001273343.1 | 2, 3 | 511 | ||
NM_014444.4 | NP_055259.2 | 2, 3 | 511 | |||
XM_011521454.2 | XP_011519756.1 | 2, 3 | 511 | |||
XM_011521455.2 | XP_011519757.1 | 3 | 409 | |||
XM_017022078.1 | XP_016877567.1 | 2, 3 | 511 | |||
GenBank mRNA | AJ249677.1 | - | 2, 3 | 217 | ||
AK026551.1 | - | 2, 3 | 498 | |||
AK027703.1 | - | 2, 3 | 469 | |||
BC009870.2 | - | 2, 3 | 436 | 3940649 | ||
BC012801.2 | - | 2, 3 | 455 | 2821891 | ||
DA396753.1 | - | 2, 3 | 511 |