Search
Search

CDS2| Gene Symbol: CDS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003818.3 | NP_003809.1 | 7 | 991 | ||
| XM_006723660.1 | XP_006723723.1 | 7 | 982 | |||
| XM_011529390.1 | XP_011527692.1 | 8 | 1314 | |||
| GenBank mRNA | AF069532.1 | - | 6 | 587 | ||
| AK001414.1 | - | 7 | 712 | |||
| AK027695.1 | - | 7 | 698 | |||
| AK091045.1 | - | 8 | 1314 | |||
| AK303426.1 | - | 3 | 265 | |||
| AK304763.1 | - | 4 | 455 | |||
| AK315489.1 | - | 7 | 972 | |||
| BC025751.1 | - | 7 | 712 | 5199748 | ||
| Y16521.1 | - | 7 | 917 | |||