Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other AURKB SIRNA Assays ›
          Gene Symbol:
          AURKB
          Gene Name:
          aurora kinase B
          Gene Aliases:
          AIK2, AIM-1, AIM1, ARK2, AurB, IPL1, PPP1R48, STK12, STK5, aurkb-sv1, aurkb-sv2
          Chromosome Location:
          Chr.17: 8204731 - 8210768 on Build GRCh38
          UniGene ID:
          Hs.442658
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 213515

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: AURKB
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001256834.2 NP_001243763.1 4 387
          NM_001284526.1 NP_001271455.1 5 463
          NM_001313950.1 NP_001300879.1 5 436
          NM_001313951.1 NP_001300880.1 5 465
          NM_001313952.1 NP_001300881.1 4 451
          NM_001313954.1 NP_001300883.1 5 460
          NM_004217.3 NP_004208.2 5 460
          NR_132731.1 4 342
          XM_011524072.2 XP_011522374.1 4 576
          XM_017025307.1 XP_016880796.1 4 503
          XM_017025309.1 XP_016880798.1 4 331
          GenBank mRNA AB011446.1 - 5 404
          AB011450.1 - 5 395
          AB519677.1 - 4 338
          AB519679.1 - 4 341
          AF004022.1 - 5 396
          AF008552.1 - 4 338
          AF015254.1 - 5 396
          AK297976.1 - 4 342
          AK314889.1 - 5 409
          AY677083.1 - 4 342
          BC000442.2 - 5 399 2820719
          BC009751.2 - 5 379 3677349
          BC013300.2 - 5 419
          BC080581.1 - 5 391 2819846
          BE514591.1 - 4 385
          BE797606.1 - 5 513
          BM905813.1 - 4 412
          BP225027.1 - 5 446
          BT019534.1 - 4 338
          BU181480.1 - 4 319
          CX781462.1 - 5 372

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          negative regulation of B cell apoptotic process
          protein phosphorylation
          spindle organization
          sister chromatid cohesion
          aging
          cell proliferation
          attachment of spindle microtubules to kinetochore
          abscission
          histone modification
          protein sumoylation
          anaphase-promoting complex-dependent catabolic process
          spindle checkpoint
          negative regulation of protein binding
          positive regulation of telomere maintenance via telomerase
          negative regulation of cytokinesis
          positive regulation of cytokinesis
          protein localization to kinetochore
          cellular response to UV
          cleavage furrow formation
          protein ubiquitination involved in ubiquitin-dependent protein catabolic process
          histone H3-S28 phosphorylation
          protein autophosphorylation
          mitotic spindle midzone assembly
          positive regulation of telomerase activity
          regulation of chromosome segregation
          regulation of signal transduction by p53 class mediator
          positive regulation of telomere capping
          protein serine/threonine kinase activity
          protein serine/threonine/tyrosine kinase activity
          protein binding
          ATP binding
          histone serine kinase activity
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0