Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PRKCI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002740.5 | NP_002731.4 | 18 | 4687 | ||
XM_017006860.1 | XP_016862349.1 | 17 | 4594 | |||
GenBank mRNA | BC042405.1 | - | 6 | 3455 |