Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMYD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006062.2 | NP_006053.2 | 13 | 1591 | ||
XM_006711918.3 | XP_006711981.1 | 13 | 1678 | |||
XM_017003163.1 | XP_016858652.1 | 12 | 3311 | |||
GenBank mRNA | BC073806.1 | - | 13 | 1567 | 6671194 | |
U50383.1 | - | 13 | 1544 |