Search
Search

MEP1B| Gene Symbol: MEP1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001308171.1 | NP_001295100.1 | 14 | 2115 | ||
| NM_005925.2 | NP_005916.2 | 14 | 2115 | |||
| XM_011526013.2 | XP_011524315.1 | 13 | 1881 | |||
| XM_011526014.2 | XP_011524316.1 | 12 | 1728 | |||
| GenBank mRNA | AY695931.1 | - | 14 | 2126 | ||
| BC136559.1 | - | 14 | 2093 | 9020549 | ||
| BC144244.1 | - | 14 | 2093 | 9052764 | ||
| X81333.1 | - | 14 | 2114 | |||