Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Pipettes and Pipette Tips
Lab Centrifuges
Ultra-Low Temperature Freezers
Spectroscopy
Beakers
PCR Equipment and Supplies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Cell Analysis
Lab Equipment
Real-Time PCR
PCR
Chromatography
Cell Culture and Transfection
DNA and RNA Extraction and Analysis
Protein Biology
Flow Cytometry
Chemicals
See all applications and techniques
Services
Custom Services
Lab Informatics
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Instrument Services
Training Services
Unity Lab Services
See all services
Help and Support
Customer Center
Contact Us
Certificates of Analysis and Conformance
Safety Data Sheets (SDS)
Manuals
How to Cite Our Products in a Paper
Instrument Support
Knowledge Base and Product FAQs
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RXFP1 SIRNA Assays ›
Gene Symbol:
RXFP1
Gene Name:
relaxin/insulin like family peptide receptor 1
Gene Aliases:
LGR7, RXFPR1
Chromosome Location:
Chr.4: 158521711 - 158653372 on Build GRCh38
UniGene ID:
Hs.591686
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
214343
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RXFP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001253727.1
NP_001240656.1
18
2403
NM_001253728.1
NP_001240657.1
17
2223
NM_001253729.1
NP_001240658.1
16
2178
NM_001253730.1
NP_001240659.1
17
2244
NM_001253732.1
NP_001240661.1
19
2406
NM_001253733.1
NP_001240662.1
18
2334
NM_021634.3
NP_067647.2
18
2322
NR_045579.1
18
2939
NR_045580.1
18
2375
NR_045581.1
18
2346
NR_045582.1
16
2283
NR_045583.1
17
2262
XM_011532174.1
XP_011530476.1
18
2369
XM_011532175.1
XP_011530477.1
17
2300
XM_011532176.1
XP_011530478.1
17
2222
XM_011532179.2
XP_011530481.1
15
1512
XM_017008517.1
XP_016864006.1
17
2297
XM_017008518.1
XP_016864007.1
18
2291
XM_017008519.1
XP_016864008.1
19
2336
XM_017008520.1
XP_016864009.1
18
2425
XM_017008521.1
XP_016864010.1
19
2255
XM_017008522.1
XP_016864011.1
18
2340
XM_017008523.1
XP_016864012.1
15
1434
XM_017008524.1
XP_016864013.1
14
1362
XM_017008525.1
XP_016864014.1
14
1366
XM_017008526.1
XP_016864015.1
13
1793
GenBank mRNA
AB208943.1
-
19
2144
AF190500.1
-
18
2059
AK122647.1
-
20
2363
AK294694.1
-
18
2167
AK295040.1
-
18
2762
AK300379.1
-
17
2118
AK307889.1
-
17
2162
AK312470.1
-
18
2166
AY899849.1
-
16
1915
AY899850.1
-
17
1960
BC112142.1
-
18
2141
8327610
BC113617.1
-
18
2144
8322669
BC143556.1
-
18
2164
BC143558.1
-
18
2197
BC143559.1
-
17
2066
BX647975.1
-
18
2260
BX647980.1
-
18
2261
BX647985.1
-
18
2289
Back To Top
More Information
Panther Classification:
Molecular Function -
G-protein coupled receptor
Gene Ontology Categories:
Function(s)
Process(es)
adenylate cyclase-modulating G-protein coupled receptor signaling pathway
G-protein coupled receptor activity
protein binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0