Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other BBS9 SIRNA Assays ›
          Gene Symbol:
          BBS9
          Gene Name:
          Bardet-Biedl syndrome 9
          Gene Aliases:
          B1, C18, D1, PTHB1
          Chromosome Location:
          Chr.7: 33129244 - 33637238 on Build GRCh38
          UniGene ID:
          Hs.372360
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 21468

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: BBS9
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001033604.1 NP_001028776.1 5, 6 963
          NM_001033605.1 NP_001028777.1 5, 6 963
          NM_014451.3 NP_055266.2 5, 6 963
          NM_198428.2 NP_940820.1 5, 6 963
          XM_005249700.3 XP_005249757.1 5, 6 971
          XM_005249701.2 XP_005249758.1 5, 6 971
          XM_011515264.2 XP_011513566.1 5, 6 971
          XM_011515265.2 XP_011513567.1 5, 6 770
          XM_011515266.2 XP_011513568.1 5, 6 971
          XM_011515267.2 XP_011513569.1 5, 6 971
          XM_011515268.1 XP_011513570.1 5, 6 971
          XM_011515269.2 XP_011513571.1 5, 6 954
          XM_011515270.2 XP_011513572.1 5, 6 971
          XM_017011990.1 XP_016867479.1 5, 6 677
          XM_017011991.1 XP_016867480.1 5, 6 679
          XM_017011992.1 XP_016867481.1 5, 6 954
          XM_017011993.1 XP_016867482.1 5, 6 954
          XM_017011994.1 XP_016867483.1 5, 6 971
          GenBank mRNA AF095771.1 - 4, 5 444
          AK057660.1 - 5, 6 941
          AK297436.1 - 2, 3 241
          AK314627.1 - 5, 6 804
          BC032715.1 - 4, 5 385
          BC103831.2 - 5, 6 583 40001794
          BX647385.1 - 5, 6 941

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          visual perception
          protein transport
          cilium assembly
          fat cell differentiation
          response to stimulus
          protein localization to cilium
          molecular_function
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-pkm9c:80/100.66.79.31:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0