Search
Search

FCRLB| Gene Symbol: FCRLB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002901.3 | NP_001002901.1 | 8 | 1373 | ||
| NM_001288829.1 | NP_001275758.1 | 6 | 1014 | |||
| NM_001288830.1 | NP_001275759.1 | 5 | 993 | |||
| NM_001288831.1 | NP_001275760.1 | 6 | 1055 | |||
| NM_001288832.1 | NP_001275761.1 | 5 | 1034 | |||
| NM_001320241.1 | NP_001307170.1 | 7 | 1187 | |||
| GenBank mRNA | AK055614.1 | - | 2 | 1367 | ||
| AK290733.1 | - | 7 | 1187 | |||
| AY670683.1 | - | 6 | 1154 | |||
| AY670684.1 | - | 6 | 1009 | |||
| AY670685.1 | - | 5 | 988 | |||
| AY670686.1 | - | 6 | 1050 | |||
| AY670687.1 | - | 5 | 1029 | |||
| AY786314.1 | - | 6 | 1139 | |||
| BC038564.2 | - | 3 | 739 | |||
| BC067080.1 | - | 2 | 1356 | |||
| DQ021957.1 | - | 8 | 1373 | |||