Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: OR5B12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004733.2 | NP_001004733.1 | 1 | 645 | ||
GenBank mRNA | BC136947.1 | - | 1 | 645 | 9020939 | |
BC136948.1 | - | 1 | 645 | 9020940 |