Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TCF7 SIRNA Assays ›
          Gene Symbol:
          TCF7
          Gene Name:
          transcription factor 7 (T-cell specific, HMG-box)
          Gene Aliases:
          TCF-1
          Chromosome Location:
          Chr.5: 134114703 - 134148229 on Build GRCh38
          UniGene ID:
          Hs.573153
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 215765

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 286.00
          Your Price
          Online offer: 277.65
          277.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TCF7
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001134851.2 NP_001128323.2 6 558
          NM_003202.3 NP_003193.2 7 1002
          NM_201632.3 NP_963963.1 6 558
          NM_201634.3 NP_963965.1 6 558
          NM_213648.3 NP_998813.1 5 792
          NR_033449.1 5 793
          XM_006714678.3 XP_006714741.1 8 1103
          XM_006714679.3 XP_006714742.1 7 1010
          XM_006714680.3 XP_006714743.1 8 1103
          XM_006714682.2 XP_006714745.1 8 1103
          XM_006714684.2 XP_006714747.1 7 1010
          XM_006714685.3 XP_006714748.1 7 648
          XM_006714686.3 XP_006714749.1 6 557
          XM_006714687.3 XP_006714750.1 6 515
          XM_011543604.2 XP_011541906.1 8 1103
          XM_011543606.1 XP_011541908.1 8 1103
          XM_011543607.2 XP_011541909.1 6 6591
          XM_011543608.2 XP_011541910.1 6 2636
          XM_011543609.2 XP_011541911.1 5 504
          XM_011543610.2 XP_011541912.1 6 636
          XM_011543611.2 XP_011541913.1 6 1530
          XM_011543612.2 XP_011541914.1 5 504
          XM_011543613.2 XP_011541915.1 5 543
          XM_017009789.1 XP_016865278.1 7 648
          XM_017009790.1 XP_016865279.1 6 1530
          GenBank mRNA AK057580.1 - 5 793
          AK093530.1 - 5 663
          AK093683.1 - 2 892
          AK131428.1 - 3 525
          AK310591.1 - 5 504
          AL832816.1 - 7 718
          AL834166.1 - 7 619
          BC048769.1 - 7 1002 6063270
          BC072023.1 - 7 937 6292932
          X59869.1 - 6 540
          X59870.1 - 6 540
          X59871.1 - 6 540
          Z47361.1 - 5 462
          Z47362.1 - 5 462

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          immune response
          neural tube development
          embryonic genitalia morphogenesis
          T cell receptor V(D)J recombination
          regulation of cell proliferation
          canonical Wnt signaling pathway involved in negative regulation of apoptotic process
          alpha-beta T cell differentiation
          embryonic digestive tract morphogenesis
          embryonic hindgut morphogenesis
          canonical Wnt signaling pathway
          cellular response to interleukin-4
          beta-catenin-TCF complex assembly
          core promoter binding
          transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
          protein binding
          beta-catenin binding
          sequence-specific DNA binding
          transcription regulatory region DNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0