Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SSNA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003731.2 | NP_003722.2 | 3 | 560 | ||
GenBank mRNA | BC000864.1 | - | 3 | 515 | 3459939 | |
Z96932.1 | - | 3 | 481 |