Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
Gene Symbol: TMEM52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178545.3 | NP_848640.1 | 4 | 348 | ||
GenBank mRNA | AJ278736.1 | - | 4 | 362 | ||
AY358544.1 | - | 1 | 508 | |||
CN263898.1 | - | 4 | 360 |