Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: KRTAP13-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181622.1 | NP_853653.1 | 1 | 209 | ||
GenBank mRNA | AB096940.1 | - | 1 | 223 | ||
BC103965.2 | - | 1 | 209 | 40014483 |