Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: EHD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014601.3 | NP_055416.2 | 5 | 1230 | ||
GenBank mRNA | AF181263.1 | - | 4 | 1018 | ||
AF454952.1 | - | 4 | 1035 | |||
AK097126.1 | - | 5 | 557 | |||
AK097228.1 | - | 4 | 549 | |||
AK293429.1 | - | 1 | 79 | |||
AK296907.1 | - | 6 | 1122 | |||
AK298067.1 | - | 4 | 693 | |||
AK315548.1 | - | 5 | 1154 | |||
AK316448.1 | - | 2 | 130 | |||
BC014445.1 | - | 5 | 1141 | 4908085 |