Search
Search

MAP1LC3C| Gene Symbol: MAP1LC3C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004343.2 | NP_001004343.1 | 4 | 328 | ||
| XM_005273139.3 | XP_005273196.1 | 5 | 356 | |||
| GenBank mRNA | AF276659.1 | - | 4 | 317 | ||
| BC127722.1 | - | 4 | 319 | 40127238 | ||
| BC132986.1 | - | 4 | 308 | 40147008 | ||
| BC132988.1 | - | 4 | 308 | 40147010 | ||
| CN260933.1 | - | 4 | 328 | |||